Novus Biologicals
Manufacturer Code:NBP297755AF700
Catalog # NB047398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SUCLA2 fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-BETA ATP-specific succinyl-CoA synthetase subunit beta beta subunit EC 6.2.1 EC 6.2.1.5 mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta MTDPS5 Renal carcinoma antigen NY-REN-39 succinate-CoA ligase ADP-forming beta subunit succinyl-CoA ligase [ADP-forming] subunit beta mitochondrial; A-SCS; ATP-specific succinyl-CoA synthetase subunit beta; ATP-specific succinyl-CoA synthetase, beta subunit; mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta; renal carcinoma antigen NY-REN-39; SCS-betaA; succinate--CoA ligase (ADP-forming); Succinate--CoA ligase [ADP-forming] subunit beta, mitochondrial; succinate-CoA ligase beta subunit; succinate-CoA ligase, ADP-forming, beta subunit; Succinyl-CoA synthetase beta-A chain
Gene Aliases: A-BETA; MTDPS5; SCS-betaA; SUCLA2
UniProt ID: (Human) Q9P2R7
Entrez Gene ID: (Human) 8803
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.