Proteintech
Manufacturer Code:109764AP150UL
Catalog # 50-172-6758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STXBP6 Fusion Protein Ag1427 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amisyn; amisyn HSPC156 STXBP6 Syntaxin binding protein 6; syntaxin binding protein 6 (amisyn); Syntaxin-binding protein 6
Gene Aliases: Amisyn; BC024598; C85317; HSPC156; STXBP6
UniProt ID: (Human) Q8NFX7
Entrez Gene ID: (Human) 29091, (Rat) 362734, (Mouse) 217517
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.