Invitrogen
Manufacturer Code:PA555549
Catalog # PIPA555549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human STXBP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Platelet Sec1 protein; Protein unc-18 homolog 3; Protein unc-18 homolog C; PSP; syntaxin 4 binding protein; Syntaxin-binding protein 3; Syntaxin-BP3 syntaxin binding protein 3 MUNC18-3 MUNC18C PSP UNC-18C platelet Sec1 protein protein unc-18 homolog 3 protein unc-18 homolog C syntaxin 4 binding protein syntaxin-binding protein 3 unc18-3; Unc-18C; Unc18-3
Gene Aliases: MUNC18-3; MUNC18C; PSP; STXBP3; UNC-18C
UniProt ID: (Human) O00186
Entrez Gene ID: (Human) 6814
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.