Proteintech
Manufacturer Code:137641AP150UL
Catalog # 50-172-8992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STXBP3 Fusion Protein Ag4747 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mammalian homolog of Unc-18c; MUNC-18-3; Munc-18c; MUNC18 3 MUNC18C Platelet Sec1 protein Protein unc 18 homolog 3 Protein unc 18 homolog C PSP STXBP3 syntaxin binding protein 3 UNC 18C Unc18 3; Platelet Sec1 protein; Protein unc-18 homolog 3; Protein unc-18 homolog C; PSP; syntaxin 4 binding protein; syntaxin binding protein 3A; Syntaxin-binding protein 3; unc-18 homolog 3; Unc-18C; Unc18-3
Gene Aliases: Munc-18c; MUNC18-3; Munc18-c; MUNC18C; PSP; STXBP3; Stxbp3a; Sxtbp3; UNC-18C; Unc18-3; Unc18c
UniProt ID: (Human) O00186, (Mouse) Q60770
Entrez Gene ID: (Human) 6814, (Rat) 114095, (Mouse) 20912
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.