Abnova
Manufacturer Code:PAB18190
Catalog # 89-119-762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human STXBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of STXBP1.
For Research Use Only
Protein Aliases: EIEE4; hUNC18; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; rbSec1; Syntaxin-binding protein 1; Unc-18A; UNC18; Unc18-1
Gene Aliases: MUNC18-1; NSEC1; P67; RBSEC1; STXBP1; UNC18; UNC18A
UniProt ID: (Human) P61764
Entrez Gene ID: (Human) 6812
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.