Proteintech
Manufacturer Code:205621AP150UL
Catalog # 50-173-2963
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EIEE4 MUNC18 1 N Sec1 p67 Protein unc 18 homolog 1 Protein unc 18 homolog A RBSEC1 STXBP1 syntaxin binding protein 1 Unc 18A UNC18 Unc18 1 UNC18A; minisatellite 10g detected by probe MMS10; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; rbSec1; Syntaxin-binding protein 1; UNC-18 homolog; unc-18-1; Unc-18A; Unc18-1
Gene Aliases: AI317162; AI326233; ANC18HA; MMS10-G; Ms10g; Munc-18a; MUNC18-1; N-sec1; NSEC1; NSEC1A; P67; Rb-sec1; RBSEC1; rbSec1A; rbSec1B; Sec1; STXBP1; Sxtbp1; UNC18; Unc18-1; UNC18A; Unc18h
UniProt ID: (Human) P61764, (Mouse) O08599, (Rat) P61765
Entrez Gene ID: (Human) 6812, (Mouse) 20910, (Rat) 25558
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.