Invitrogen
Manufacturer Code:PA570522
Catalog # PIPA570522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human STX7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Syntaxin-7; Syntaxin-7 STX7 Syn7; syntaxin-7-like; syntaxin-7-like protein
Gene Aliases: AI315064; AI317144; AW107239; STX7; Syn7
UniProt ID: (Human) O15400, (Bovine) Q3ZBT5, (Rat) O70257
Entrez Gene ID: (Dog) 483987, (Horse) 100073104, (Rabbit) 100343660, (Human) 8417, (Bovine) 507031, (Rat) 60466, (Mouse) 53331
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.