Invitrogen
Manufacturer Code:OSS00054W
Catalog # PIOSS00054W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from mouse Syntaxin-7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Syntaxin-7; Syntaxin-7 STX7 Syn7
Gene Aliases: AI315064; AI317144; AW107239; Stx7; Syn7
UniProt ID: (Rat) O70257
Entrez Gene ID: (Rat) 60466, (Mouse) 53331
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.