Invitrogen
Manufacturer Code:PA571665
Catalog # PIPA571665
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 180-209 amino acids from the Central region of human STX5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SED5 STX5A syntaxin 5A syntaxin-5; syntaxin 5A; Syntaxin-5
Gene Aliases: 0610031F24Rik; D19Ertd627e; SED5; STX5; STX5A
UniProt ID: (Human) Q13190, (Mouse) Q8K1E0
Entrez Gene ID: (Human) 6811, (Mouse) 56389
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.