Novus Biologicals
Manufacturer Code:NBP293340100UL
Catalog # NB028564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human STX18 (NP_058626.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 9 protein; Cell growth-inhibiting gene 9 protein DKFZp686O15149 syntaxin 18 syntaxin-18 Ufe1; Syntaxin-18
Gene Aliases: GIG9; STX18; Ufe1
UniProt ID: (Human) Q9P2W9
Entrez Gene ID: (Human) 53407
Molecular Function:
SNARE protein
membrane traffic protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.