Invitrogen
Manufacturer Code:PA518011
Catalog # PIPA518011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DAFISENGWVEDY) corresponding to the C-terminus amino acids of STUB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-7; Carboxy terminus of Hsp70-interacting protein; CLL-associated antigen KW-8; E3 ubiquitin-protein ligase CHIP; heat shock protein A binding protein 2 (c-terminal); RING-type E3 ubiquitin transferase CHIP; serologically defined colon cancer antigen 7; STIP1 homology and U box-containing protein 1; STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase; STUB1 CHIP STIP1 homology and U-Box containing protein 1 HSPABP2 NY-CO-7 SDCCAG7 serologically defined colon cancer antigen 7 carboxy terminus of Hsp70p-interacting protein heat shock protein A binding protein 2 (c-terminal) LA16c-313D11.6 UBOX1
Gene Aliases: CHIP; HSPABP2; NY-CO-7; PP1131; SCAR16; SDCCAG7; STUB1; UBOX1
UniProt ID: (Human) Q9UNE7
Entrez Gene ID: (Human) 10273
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.