Proteintech
Manufacturer Code:153231AP150UL
Catalog # 50-173-0149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STT3B Fusion Protein Ag2039 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B6dom1 antigen; dolichyl-diphosphooligosaccharide protein glycotransferase; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3B; homolog of yeast STT3; Oligosaccharyl transferase subunit STT3B; SIMP STT3 B STT3B; Source of immunodominant MHC-associated peptides; Source of immunodominant MHC-associated peptides homolog; STT3, subunit of the oligosaccharyltransferase complex, homolog B; STT3-B; STT3B, subunit of the oligosaccharyltransferase complex (catalytic)
Gene Aliases: 1300006C19Rik; CDG1X; RGD1311563; SIMP; STT3-B; STT3B
UniProt ID: (Human) Q8TCJ2, (Mouse) Q3TDQ1
Entrez Gene ID: (Human) 201595, (Rat) 363160, (Mouse) 68292
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.