Abnova
Manufacturer Code:PAB21324
Catalog # 89-122-815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human STRBP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant STRBP.
For Research Use Only
Protein Aliases: DKFZp434N214; epididymis luminal protein 162; FLJ11307; FLJ14223; FLJ14984; ILF3L; MGC21529; MGC3405; p74; Spermatid perinuclear RNA-binding protein; SPNR
Gene Aliases: HEL162; ILF3L; p74; SPNR; STRBP
UniProt ID: (Human) Q96SI9
Entrez Gene ID: (Human) 55342
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.