Proteintech
Manufacturer Code:173621AP150UL
Catalog # 50-173-1531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STRBP Fusion Protein Ag10720 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 74 kDa double-stranded RNA-binding protein; double-stranded RNA-binding protein p74; epididymis luminal protein 162; ILF3L p74 SPNR STRBP; p74; Spermatid perinuclear RNA-binding protein
Gene Aliases: 6430510M02Rik; AI852513; C230082I21Rik; C86322; HEL162; ILF3L; p74; SPNR; STRBP
UniProt ID: (Human) Q96SI9, (Mouse) Q91WM1
Entrez Gene ID: (Human) 55342, (Rat) 84476, (Mouse) 20744
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.