Invitrogen
Manufacturer Code:MA524722
Catalog # PIMA524722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 526CT11.1.5 |
Immunogen | KLH conjugated synthetic peptide between 31-59 amino acids from human STRA8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EMBL:AAP471631}; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8 homolog; Stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; Stimulated by retinoic acid gene 8 protein homolog STRA8 {ECO:0000312; STRA8
Gene Aliases: STRA8
UniProt ID: (Human) Q7Z7C7, (Mouse) P70278
Entrez Gene ID: (Human) 346673, (Mouse) 20899
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.