Invitrogen
Manufacturer Code:PA5115977
Catalog # PIPA5115977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Stra8 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RGD1562852 stimulated by retinoic acid 8 stimulated by retinoic acid 8 homolog stimulated by retinoic acid gene 8 stimulated by retinoic acid gene 8 homolog stimulated by retinoic acid gene 8 protein Stimulated by retinoic acid gene 8 protein homolog STRA8; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; Stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; STRA8
Gene Aliases: RGD1562852; STRA8
UniProt ID: (Human) Q7Z7C7, (Mouse) P70278
Entrez Gene ID: (Human) 346673, (Mouse) 20899, (Rat) 500079
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.