Novus Biologicals
Manufacturer Code:NBP159711
Catalog # NBP159711
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to STRA6(stimulated by retinoic acid gene 6 homolog (mouse)) The peptide sequence was selected from the N terminal of STRA6 (NP_071764). Peptide sequence MSSQPAGNQTSPGATEDYSYGSWYIDEPQGGEELQPEGEVPSCHTSIPPG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ12541 MCOPS9 stimulated by retinoic acid gene 6 homolog (mouse) stimulated by retinoic acid gene 6 protein homolog; Receptor for retinol uptake STRA6; retinol binding protein 4 receptor; Retinol-binding protein receptor STRA6; stimulated by retinoic acid 6 homolog; stimulated by retinoic acid gene 6 homolog; Stimulated by retinoic acid gene 6 protein homolog
Gene Aliases: MCOPCB8; MCOPS9; PP14296; STRA6; UNQ3126/PRO10282/PRO19578
UniProt ID: (Human) Q9BX79
Entrez Gene ID: (Human) 64220
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.