Proteintech
Manufacturer Code:128621AP150UL
Catalog # 50-172-8309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STOML1 Fusion Protein Ag3978 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EPB72 like protein 1 hUNC 24 Protein unc 24 homolog SLP 1 SLP1 stomatin (EPB72) like 1 Stomatin like protein 1 Stomatin related protein STOML1 STORP UNC24; EPB72-like 1; EPB72-like protein 1; Protein unc-24 homolog; SLP-1; stomatin (EBP72)-like 1; stomatin (EPB72)-like 1; stomatin-like 1; Stomatin-like protein 1; Stomatin-related protein; STORP
Gene Aliases: 1810015E19Rik; AI847431; hUNC-24; MSTP019; RGD1559463; SLP-1; SLP1; STOML1; STORP; UNC24; WPB72
UniProt ID: (Human) Q9UBI4, (Mouse) Q8CI66
Entrez Gene ID: (Human) 9399, (Rat) 300748, (Mouse) 69106
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.