Invitrogen
Manufacturer Code:PA5110443
Catalog # PIPA5110443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 55-288 of human STOM (NP_004090.4). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BND7 EPB7 Epb7.2 Epb72 Erythrocyte band 7 integral membrane protein Protein 7.2b STOM Stomatin; Erythrocyte band 7 integral membrane protein; Erythrocyte membrane protein band 7.2; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte protein band 7.2; erythrocyte protein band 7.2; protein 7.2b; erythrocyte surface protein band 7.2; Protein 7.2b; Stomatin
Gene Aliases: BND7; EPB7; Epb7.2; EPB72; STOM
UniProt ID: (Human) P27105, (Mouse) P54116
Entrez Gene ID: (Human) 2040, (Rat) 296655, (Mouse) 13830
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.