Abnova
Manufacturer Code:H00002040A01
Catalog # 89-104-444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 59 a.a. &bsim 142 a.a) partial recombinant protein with GST tag.,STOM (NP_004090 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant STOM.
For Research Use Only
Protein Aliases: BND7; EPB7; EPB72; Erythrocyte band 7 integral membrane protein; Erythrocyte membrane protein band 7.2; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte surface protein band 7.2; Protein 7.2b; Stomatin
Gene Aliases: BND7; EPB7; EPB72; STOM
UniProt ID: (Human) P27105
Entrez Gene ID: (Human) 2040
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.