Proteintech
Manufacturer Code:120271AP150UL
Catalog # 50-172-7655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STMN4 Fusion Protein Ag2657 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RB3; RB3 Stathmin 4 stathmin like 4 Stathmin like protein B3 STMN4; Stathmin-4; stathmin-like 4; Stathmin-like protein B3; stathmin-like protein RB3; stathmin-like-protein RB3
Gene Aliases: Lagl; R75359; RB3; STMN4
UniProt ID: (Human) Q9H169, (Rat) P63043, (Mouse) P63042
Entrez Gene ID: (Human) 81551, (Rat) 79423, (Mouse) 56471
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.