Invitrogen
Manufacturer Code:PA515179
Catalog # PIPA515179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 346-376 amino acids from the central region of human SPAK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCHT; kidney-specific SPAK; proline-alanine-rich STE20-related kinase; serine threonine kinase 39 (STE20/SPS1 homolog, yeast); serine/threonine kinase 39, STE20/SPS1 homolog; Serine/threonine-protein kinase 39; small intestine SPAK-like kinase; Ste-20 related kinase; Ste-20-related kinase; Ste20-like protein kinase; STE20/SPS1 homolog; STE20/SPS1-related proline-alanine rich protein kinase Ste-20 related kinase Serine/threonine-protein kinase 39 DCHT; STE20/SPS1-related proline-alanine-rich protein kinase
Gene Aliases: AW227544; AW556857; DCHT; PASK; RF005; Rnl5; SPAK; STK39
UniProt ID: (Human) Q9UEW8, (Mouse) Q9Z1W9
Entrez Gene ID: (Human) 27347, (Mouse) 53416
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.