Invitrogen
Manufacturer Code:PA514803
Catalog # PIPA514803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 408-437 amino acids from the C-terminal region of human STK38L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NDR2 protein kinase; nuclear Dbf2-related 2; Nuclear Dbf2-related kinase 2; Serine/threonine-protein kinase 38-like; Serine/threonine-protein kinase 38-like NDR2 protein kinase Nuclear Dbf2-related kinase 2 STK38L KIAA0965 NDR2; STK38L
Gene Aliases: KIAA0965; NDR2; STK38L
UniProt ID: (Human) Q9Y2H1
Entrez Gene ID: (Human) 23012
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.