Proteintech
Manufacturer Code:126971AP150UL
Catalog # 50-172-8182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STK38L Fusion Protein Ag3413 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0965 NDR2 NDR2 protein kinase Nuclear Dbf2 related kinase 2 STK38L; NDR2 protein kinase; nuclear Dbf2-related 2; Nuclear Dbf2-related kinase 2; serine/threonine kinase 38-like; Serine/threonine-protein kinase 38-like; STK38L
Gene Aliases: 4930473A22Rik; B230328I19; KIAA0965; NDR2; Ndr54; RGD1564793; STK38L
UniProt ID: (Human) Q9Y2H1, (Mouse) Q7TSE6
Entrez Gene ID: (Human) 23012, (Mouse) 232533, (Rat) 691337
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.