Proteintech
Manufacturer Code:258211AP150UL
Catalog # 50-173-5087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STK25 Fusion Protein Ag22964 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: serine/threonine kinase 25 (Ste20, yeast homolog); Serine/threonine-protein kinase 25; SOK 1 SOK1 Ste20 like kinase STK25 YSK1; SOK-1; Ste20, yeast homolog; Ste20-like kinase; ste20/oxidant stress response kinase 1; Sterile 20/oxidant stress-response kinase 1; Yeast Sps1/Ste20-related kinase 1
Gene Aliases: 1500019J11Rik; AU018434; C86992; SOK1; Ste20-like; STK25; YSK1
UniProt ID: (Human) O00506, (Mouse) Q9Z2W1
Entrez Gene ID: (Human) 10494, (Mouse) 59041
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.