Invitrogen
Manufacturer Code:PA570420
Catalog # PIPA570420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of rat STING |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum IFN stimulator; Endoplasmic reticulum interferon stimulator; ERIS; ERIS MITA MPYS NET23 STING N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner endoplasmic reticulum IFN stimulator endoplasmic reticulum interferon stimulator hMITA hSTING mediator of IRF3 activation mitochondrial mediator of IRF3 activation stimulator of interferon genes protein; hMITA; hSTING; Mediator of IRF3 activation; mitochondrial mediator of IRF3 activation; mitochondrial transmembrane protein 173; MMITA; mSTING; N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner; poSTING; rSTING; Stimulator of interferon genes protein; STING; Transmembrane protein 173
Gene Aliases: 2610307O08Rik; ERIS; hMITA; hSTING; MITA; MPYS; NET23; RGD1562552; SAVI; STING; STING1; TMEM173
UniProt ID: (Pig) B8XX90, (Bovine) Q2KI99, (Human) Q86WV6, (Mouse) Q3TBT3, (Rat) F1M391
Entrez Gene ID: (Pig) 100217389, (Dog) 606815, (Rabbit) 100349135, (Horse) 100062046, (Bovine) 533661, (Human) 340061, (Mouse) 72512, (Rat) 498840, (Guinea pig) 100732510
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.