Invitrogen
Manufacturer Code:PA5116052
Catalog # PIPA5116052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TMEM173/STING corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610307O08Rik endoplasmic reticulum IFN stimulator endoplasmic reticulum interferon stimulator Eris hMITA hSTING Mediator of IRF3 activation MITA mitochondrial mediator of IRF3 activation mitochondrial transmembrane protein 173 MMITA Mpys mSTING NET23 N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner RGD1562552 rSTING SAVI Stimulator of interferon genes protein STING tm173 Tmem173 transmembrane protein 173; endoplasmic reticulum IFN stimulator; Endoplasmic reticulum interferon stimulator; ERIS; hMITA; hSTING; Mediator of IRF3 activation; mitochondrial mediator of IRF3 activation; mitochondrial transmembrane protein 173; MMITA; mSTING; N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner; rSTING; Stimulator of interferon genes protein; Transmembrane protein 173
Gene Aliases: 2610307O08Rik; ERIS; hMITA; hSTING; MITA; MPYS; NET23; RGD1562552; SAVI; STING; STING1; TMEM173
UniProt ID: (Human) Q86WV6, (Mouse) Q3TBT3, (Rat) F1M391
Entrez Gene ID: (Human) 340061, (Mouse) 72512, (Rat) 498840
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.