Invitrogen
Manufacturer Code:MA1158
Catalog # PIMA1158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 23E5 |
Immunogen | A synthetic peptide of STEP46 (amino acids 57-74: MCTPGCNEEGFGYLVSPR) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Neural-specific protein-tyrosine phosphatase; protein-tyrosine phosphatase striatum-enriched; protein-tyrosine-phosphatase non-receptor 5; STEP; STEP Striatum-enriched protein-tyrosine phosphatase Neural-specific protein-tyrosine phosphatase PTPN5 PTN5 Tyrosine-protein phosphatase non-receptor type 5; Striatum-enriched protein-tyrosine phosphatase; Tyrosine-protein phosphatase non-receptor type 5
Gene Aliases: Ptpn5; STEP
UniProt ID: (Rat) P35234
Entrez Gene ID: (Rat) 29644
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.