Invitrogen
Manufacturer Code:MA116746
Catalog # AMPA6746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 23E5 |
Immunogen | A synthetic peptide of STEP46 (amino acids 57-74: MCTPGCNEEGFGYLVSPR) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Neural-specific protein-tyrosine phosphatase; protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched); protein-tyrosine phosphatase striatum-enriched; protein-tyrosine-phosphatase non-receptor 5; STEP; striatal-enriched protein tyrosine phosphatase; Striatum-enriched protein-tyrosine phosphatase; Tyrosine-protein phosphatase non-receptor type 5
Gene Aliases: PTPN5; PTPSTEP; STEP; STEP61
UniProt ID: (Human) P54829, (Mouse) Q64694, (Rat) P35234
Entrez Gene ID: (Human) 84867, (Mouse) 19259, (Rat) 29644
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.