Invitrogen
Manufacturer Code:PA515531
Catalog # PIPA515531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 168-198 amino acids from the N-terminal region of human STEP / PTPN5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ14427 Neural specific protein tyrosine phosphatase Protein tyrosine phosphatase non receptor type 5 (striatum enriched) Protein tyrosine phosphatase non receptor type 5 Protein tyrosine phosphatase striatum enriched PTN5 PTP STEP PTPN 5 PTPSTEP Tyrosine protein phosphatase non receptor type 5; Neural-specific protein-tyrosine phosphatase; protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched); protein-tyrosine phosphatase striatum-enriched; STEP; striatal-enriched protein tyrosine phosphatase; Striatum-enriched protein-tyrosine phosphatase; Tyrosine-protein phosphatase non-receptor type 5
Gene Aliases: PTPN5; PTPSTEP; STEP; STEP61
UniProt ID: (Human) P54829, (Mouse) Q64694
Entrez Gene ID: (Human) 84867, (Mouse) 19259
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.