Invitrogen
Manufacturer Code:PA534225
Catalog # PIPA534225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from N-Terminus of human STEAP4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Metalloreductase STEAP4; six transmembrane prostate protein 2; Six-transmembrane epithelial antigen of prostate 4; SixTransMembrane protein of prostate 2; STEAP family member 4; STEAP4 STEAP family member 4 Metalloreductase STEAP4 STAMP2 TIARP TNFAIP9; Tumor necrosis factor, alpha-induced protein 9; tumor necrosis-alpha-induced adipose-related protein
Gene Aliases: STAMP2; STEAP4; TIARP; TNFAIP9
UniProt ID: (Human) Q687X5
Entrez Gene ID: (Human) 79689
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.