Abnova
Manufacturer Code:PAB7348
Catalog # 89-133-169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human STEAP4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of STEAP4.
For Research Use Only
Protein Aliases: DKFZp666D049; FLJ23153; Metalloreductase STEAP4; six transmembrane prostate protein 2; Six-transmembrane epithelial antigen of prostate 4; SixTransMembrane protein of prostate 2; STAMP2; STEAP family member 4; TIARP; TNFAIP9; Tumor necrosis factor, alpha-induced protein 9; tumor necrosis-alpha-induced adipose-related protein
Gene Aliases: STAMP2; STEAP4; TIARP; TNFAIP9
UniProt ID: (Human) Q687X5
Entrez Gene ID: (Human) 79689
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.