Proteintech
Manufacturer Code:119441AP150UL
Catalog # 50-172-7580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STEAP4 Fusion Protein Ag2545 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ab1-219; dudulin 4; Dudulin-4; Metalloreductase STEAP4; Metalloreductase STEAP4 STAMP2 STEAP family member 4 STEAP4 TIARP TNFAIP9; six transmembrane prostate protein 2; Six-transmembrane epithelial antigen of prostate 4; SixTransMembrane protein of prostate 2; STEAP family member 4; Tnfa-induced adipose-related protein; Tumor necrosis factor, alpha-induced protein 9; Tumor necrosis factor-alpha-induced adipose-related protein; tumor necrosis-alpha-induced adipose-related protein
Gene Aliases: 1110021O17Rik; AI481214; STAMP2; STEAP4; TIARP; TNFAIP9
UniProt ID: (Human) Q687X5, (Rat) Q4V8K1, (Mouse) Q923B6
Entrez Gene ID: (Human) 79689, (Rat) 499991, (Mouse) 117167
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.