Novus Biologicals
Manufacturer Code:NBP295115100UL
Catalog # NB024392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human STEAP3 (NP_060704.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dudlin 2 dudlin-2 dudulin 2 dudulin-2 Dudulin-21010001D01Rik EC 1.16.1.- hpHyde hTSAP6 metalloreductase STEAP3 pHyde six transmembrane prostate protein 3 Six-transmembrane epithelial antigen of prostate 3 STEAP family member 3 TSAP6STMP3 tumor suppressor pHyde Tumor suppressor-activated pathway protein 6; Dudulin-2; hpHyde; hTSAP6; Metalloreductase STEAP3; pHyde; Six-transmembrane epithelial antigen of prostate 3; STEAP family member 3, metalloreductase; tumor suppressor pHyde; Tumor suppressor-activated pathway protein 6
Gene Aliases: AHMIO2; dudlin-2; dudulin-2; pHyde; STEAP3; STMP3; TSAP6
UniProt ID: (Human) Q658P3
Entrez Gene ID: (Human) 55240
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.