Invitrogen
Manufacturer Code:PA587478
Catalog # PIPA587478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-180 of human STEAP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dudulin 2; Dudulin-2; hpHyde; hTSAP6; Metalloreductase STEAP3; pHyde; Protein nm1054; Six-transmembrane epithelial antigen of prostate 3; STEAP family member 3, metalloreductase; tumor suppressor pHyde; Tumor suppressor-activated pathway protein 6
Gene Aliases: 1010001D01Rik; AHMIO2; dudlin-2; dudulin-2; pHyde; STEAP3; STMP3; TSAP6
UniProt ID: (Human) Q658P3, (Mouse) Q8CI59
Entrez Gene ID: (Human) 55240, (Mouse) 68428
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.