Proteintech
Manufacturer Code:171861AP150UL
Catalog # 50-173-1430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STEAP3 Fusion Protein Ag10970 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dudlin 2 Dudulin 2 hpHyde hTSAP6 Metalloreductase STEAP3 pHyde STEAP family member 3 STEAP3 STMP3 TSAP6; dudulin 2; Dudulin-2; hpHyde; hTSAP6; Metalloreductase STEAP3; pHyde; Protein nm1054; Six-transmembrane epithelial antigen of prostate 3; STEAP family member 3, metalloreductase; tumor suppressor pHyde; Tumor suppressor-activated pathway protein 6
Gene Aliases: 1010001D01Rik; AHMIO2; dudlin-2; dudulin-2; pHyde; STEAP3; STMP3; TSAP6
UniProt ID: (Human) Q658P3, (Mouse) Q8CI59, (Rat) Q5RKL5
Entrez Gene ID: (Human) 55240, (Mouse) 68428, (Rat) 170824
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.