Invitrogen
Manufacturer Code:PA575929
Catalog # PIPA575929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 440-485 of Human STEAP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Metalloreductase STEAP2; Metalloreductase STEAP2 Prostate cancer-associated protein 1 Protein up-regulated in metastatic prostate cancer PUMPCn Six-transmembrane epithelial antigen of prostate 2 SixTransMembrane protein of prostate 1 STEAP2 PCANAP1 STAMP1 UNQ6507/PRO23203; prostate cancer associated protein 1; Prostate cancer-associated protein 1; Protein up-regulated in metastatic prostate cancer; protein upregulated in metastatic prostate cancer; PUMPCn; six transmembrane epithelial antigen of prostate 2; six transmembrane epithelial antigen of the prostate 2; Six-transmembrane epithelial antigen of prostate 2; SixTransMembrane Protein of Prostate 1; STEAP family member 2, metalloreductase
Gene Aliases: 4921538B17Rik; AI930049; AW045895; IPCA-1; IPCA1; PCANAP1; PUMPCn; STAMP1; STEAP2; STMP; UNQ6507/PRO23203
UniProt ID: (Human) Q8NFT2, (Mouse) Q8BWB6
Entrez Gene ID: (Human) 261729, (Rat) 312052, (Mouse) 74051
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.