Abnova
Manufacturer Code:H00261729K
Catalog # 89-204-896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human STEAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human STEAP2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: IPCA1 PCANAP1 PUMPCn STAMP1 STMP; Metalloreductase STEAP2; prostate cancer associated protein 1; Prostate cancer-associated protein 1; Protein up-regulated in metastatic prostate cancer; protein upregulated in metastatic prostate cancer; PUMPCn; six transmembrane epithelial antigen of prostate 2; six transmembrane epithelial antigen of the prostate 2; Six-transmembrane epithelial antigen of prostate 2; SixTransMembrane Protein of Prostate 1; STEAP family member 2, metalloreductase
Gene Aliases: IPCA1; PCANAP1; PUMPCn; STAMP1; STEAP2; STMP; UNQ6507/PRO23203
UniProt ID: (Human) Q8NFT2
Entrez Gene ID: (Human) 261729
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.