Novus Biologicals
Manufacturer Code:NBP107094AF488
Catalog # NBP107094X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | J2D2 |
Immunogen | Recombinant human STEAP1 (1-70aa) purified from E. coli (NP_036581). |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.16.1.- PRSS24MGC19484 six transmembrane epithelial antigen of the prostate six transmembrane epithelial antigen of the prostate 1 Six-transmembrane epithelial antigen of prostate 1 STEAPmetalloreductase STEAP1; Metalloreductase STEAP1; Six-transmembrane epithelial antigen of prostate 1
Gene Aliases: PRSS24; STEAP; STEAP1
UniProt ID: (Human) Q9UHE8
Entrez Gene ID: (Human) 26872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.