Invitrogen
Manufacturer Code:PA520404
Catalog # PIPA520404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide from near the center of human STEAP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Metalloreductase STEAP1; Six-transmembrane epithelial antigen of prostate 1; tcag7.1170 PRSS24 STEAP metalloreductase STEAP1 six-transmembrane epithelial antigen of prostate 1
Gene Aliases: 2410007B19Rik; PRSS24; STEAP; STEAP1
UniProt ID: (Human) Q9UHE8, (Mouse) Q9CWR7
Entrez Gene ID: (Human) 26872, (Rat) 297738, (Mouse) 70358
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.