R&D Systems
Manufacturer Code:AF5587SP
Catalog # AF5587SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human STEAP1 Met1-His72 Accession # Q9UHE8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.16.1.- PRSS24 PRSS24MGC19484 six transmembrane epithelial antigen of the prostate six transmembrane epithelial antigen of the prostate 1 Six-transmembrane epithelial antigen of prostate 1 STEAPmetalloreductase STEAP1; Metalloreductase STEAP1; Six-transmembrane epithelial antigen of prostate 1
Gene Aliases: PRSS24; STEAP; STEAP1
UniProt ID: (Human) Q9UHE8
Entrez Gene ID: (Human) 26872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.