Invitrogen
Manufacturer Code:PA511501
Catalog # PIPA511501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 16-46 amino acids from the N-terminal region of human Stanniocalcin-2 (STC2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Stanniocalcin-2; Stanniocalcin-2 STC-2 Stanniocalcin-related protein STCRP STC-related protein; Stanniocalcin-related protein; STC-2; STC-related protein
Gene Aliases: AW125853; mustc2; STC-2; STC2; Stc2l; STCRP
UniProt ID: (Human) O76061, (Mouse) O88452
Entrez Gene ID: (Human) 8614, (Mouse) 20856
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.