Invitrogen
Manufacturer Code:PA5106379
Catalog # PIPA5106379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Stanniocalcin 2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW125853 mustc2 stanniocalcin 2 stanniocalcin-2 Stanniocalcin-related protein STC2 STC-2 Stc2l STC-related protein STCRP; Stanniocalcin-2; Stanniocalcin-related protein; STC-2; STC-related protein
Gene Aliases: AW125853; mustc2; STC-2; STC2; Stc2l; STCRP
UniProt ID: (Human) O76061, (Mouse) O88452
Entrez Gene ID: (Human) 8614, (Mouse) 20856
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.