Invitrogen
Manufacturer Code:PA542104
Catalog # PIPA542104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human STC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Stanniocalcin 1 STC; Stanniocalcin-1; STC-1
Gene Aliases: STC; STC1
UniProt ID: (Bovine) Q9N0T1, (Human) P52823, (Mouse) O55183, (Rat) P97574
Entrez Gene ID: (Rabbit) 100353589, (Bovine) 338078, (Human) 6781, (Dog) 486112, (Horse) 100054071, (Pig) 100125345, (Guinea pig) 100731077, (Mouse) 20855, (Rat) 81801
Molecular Function:
peptide hormone
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.