Novus Biologicals
Manufacturer Code:NBP224826
Catalog # NBP224826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The STAT6 antibody was developed against a synthetic peptide corresponding to amino acids 808-840 of human Signal Transducer and Activator of Transcription 6 . This antibody will cross-react with mouse and rat based on NCBI BLAST. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D12S1644 EC 2.4.1.227 EC 2.7.7.6 IL-4 Stat IL-4-STAT signal transducer and activator of transcription 6 signal transducer and activator of transcription 6 interleukin-4 induced STAT interleukin-4 induced STAT interleukin4-induced STAT6B STAT6C transcription factor IL-4 STAT; IL-4 Stat; Signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; STAT, interleukin4-induced; transcription factor IL-4 STAT
Gene Aliases: D12S1644; IL-4-STAT; STAT6; STAT6B; STAT6C
UniProt ID: (Human) P42226
Entrez Gene ID: (Human) 6778
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.