OriGene
Manufacturer Code:TA500177
Catalog # 50-167-1416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI21E7 |
Immunogen | Recombit protein expressed in E.coli corresponding to amino acids 154-175 of human stat3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acute-phase response factor; ADMIO ADMIO1 APRF HIES; DNA-binding protein APRF; Signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor)
Gene Aliases: ADMIO; ADMIO1; APRF; HIES; STAT3
UniProt ID: (Human) P40763
Entrez Gene ID: (Human) 6774
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.