Invitrogen
Manufacturer Code:PA585199
Catalog # PIPA585199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human STAT3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acute phase response factor; Acute-phase response factor; APRF antibody FLJ20882 antibody HIES antibody MGC16063 antibody STAT3 antibody acute-phase response factor antibody DNA-binding protein APRF antibody signal transducer and activator of transcription 3 antibody signal transducer and activator of transcription 3 (acute-phase response factor) antibody; DNA-binding protein APRF; Signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor)
Gene Aliases: 1110034C02Rik; ADMIO; ADMIO1; APRF; AW109958; HIES; STAT3
UniProt ID: (Human) P40763, (Rat) P52631, (Mouse) P42227
Entrez Gene ID: (Human) 6774, (Rat) 25125, (Mouse) 20848
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.