R&D Systems
Manufacturer Code:AF1799SP
Catalog # AF1799SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse STAT3 Arg688-Met770 Accession # U06922 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Acute-phase response factor; Acute-phase response factor APRFMGC16063 DNA-binding protein APRF FLJ20882 HIES signal transducer and activator of transcription 3 signal transducer and activator of transcription 3 (acute-phase responsefactor); DNA-binding protein APRF; Signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor)
Gene Aliases: ADMIO; ADMIO1; APRF; HIES; STAT3
UniProt ID: (Human) P40763
Entrez Gene ID: (Human) 6774
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.