Proteintech
Manufacturer Code:510762AP150UL
Catalog # 50-173-5894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acute phase response factor; Acute phase response factor APRF HIES STAT3; Acute-phase response factor; DNA-binding protein APRF; Signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor)
Gene Aliases: 1110034C02Rik; ADMIO; ADMIO1; APRF; AW109958; HIES; STAT3
UniProt ID: (Human) P40763, (Mouse) P42227
Entrez Gene ID: (Human) 6774, (Mouse) 20848
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.