R&D Systems
Manufacturer Code:MAB16662100
Catalog # MAB16662100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 967425 |
Immunogen | <em>E. coli</em>-derived recombinant human STAT2 Gln679-Phe851 Accession # P52630 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: interferon alpha induced transcriptional activator; interferon alpha induced transcriptional activator ISGF-3 MGC59816 p113 signal transducer and activator of transcription 2 signal transducer and activator of transcription 2 113kD signal transducer and activator of transcription 2 113kDa STAT113; p113; Signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa
Gene Aliases: IMD44; ISGF-3; P113; STAT113; STAT2
UniProt ID: (Human) B4DLC7
Entrez Gene ID: (Human) 6773
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.